Documenti fondamentali
EP0030129610
Eppendorf twin.tec® PCR Plates 96
Semi-Skirted, Forensic DNA Grade, working volume 250 μL, colorless, pack of 10 (individually wrapped)
Scegli un formato
10 EA
CHF 82.50
CHF 82.50
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 EA
CHF 82.50
About This Item
Codice UNSPSC:
41106300
NACRES:
NB.22
CHF 82.50
Check Cart for Availability
Materiali
colorless
colorless wells
Dimensione pozzetto
96
Caratteristiche
skirt
Forensic DNA Grade
Semi-Skirted
Skirt (Semi skirted)
Confezionamento
pack of 10 (individually wrapped)
Produttore/marchio commerciale
Eppendorf® 0030129610 (Global)
tecniche
PCR: suitable
Misura
96 well
Volume d’esercizio
250 μL
Descrizione generale
Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.
Applicazioni
PCR
Caratteristiche e vantaggi
  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Note legali
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)