Documenti fondamentali
EP0030128940
Eppendorf® Twin tec PCR plates, 384 wells
yellow polypropylene plate, conical bottom colorless wells, pkg of 300 ea
Scegli un formato
300 EA
CHF 2’620.00
CHF 2’620.00
Spedizione prevista il20 maggio 2025
Scegli un formato
Cambia visualizzazione
300 EA
CHF 2’620.00
About This Item
Codice UNSPSC:
41106300
NACRES:
NB.15
CHF 2’620.00
Spedizione prevista il20 maggio 2025
Materiali
conical bottom colorless wells
yellow polypropylene plate
Caratteristiche
skirt (stackable)
Confezionamento
pkg of 300 ea
Produttore/marchio commerciale
Eppendorf® 0030128940
tecniche
PCR: suitable
Volume massimo
-45 μL
Misura
384 wells
Volume d’esercizio
-40 μL
Compatibilità
suitable for PCR
Tipo di binding
low binding surface
Descrizione generale
The Eppendorf “twin.tec” PCR plates are designed for single-use PCR applications. They have a rigid polycarbonate frame that retains its shape even when exposed to high temperatures during the PCR cycle. The plate is designed to maintain dimensional stability, providing support for both the snug fit and sealing functions. Additionally, the raised rims of the plate further enhance effective sealing by forming a highly secure seal with the sealing material.
Applicazioni
“Eppendorf” Twin tec PCR plates are suitable for:
  • Standard PCR
  • Real-time PCR
  • High Throughput PCR
  • Small Volume Sample Handling
Caratteristiche e vantaggi
  • Eppendorf “twin.tec” PCR plate has a precise fit to the PCR cycler and a sturdy base enables tight sealing.
  • It is free of prevalent PCR contaminants such as DNA, DNAse, RNAse, and PCR inhibitors.
  • Skirt is designed to optimize automation and barcoding.
  • The eight holes in the skirt facilitate easy positioning and removal of the plate from the thermal cycler block.
  • Idea for use in 384-well thermocyclers.
  • Maximum volume per well is 40 µL.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)