Documenti fondamentali
EP0030048130
Eppendorf® Varitips P
pkg of 100 pieces
Scegli un formato
100 EA
CHF 142.00
CHF 142.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
100 EA
CHF 142.00
About This Item
Codice UNSPSC:
41121600
NACRES:
NB.02
CHF 142.00
Check Cart for Availability
Sterilità
non-sterile
Confezionamento
pack of 100 ea
pkg of 100 pieces
Produttore/marchio commerciale
Eppendorf® 0030048130
Caratteristiche e vantaggi
  • Use Varitips P for aspirating 1 mL to 10 mL from beakers using the positive displacement principle
  • Use Varitips S in combination with the Eppendorf Maxitip to aspirate between 2.5 mL and 10 mL of liquid from tall, narrow-necked vessels using the air cushion principle
  • Valve for Varitips ensures drip-free dosing of liquids with high vapor pressure
  • The graduation on Varitips ensures accurate dosing of non-aqueous liquids
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)