CLS766116
Pyrex® pennyhead stopper
stopper joint: 16
Sinonimo/i:
glass stopper, glass stoppers, stoppers, Corning® 7660
About This Item
Prodotti consigliati
Materiali
glass
Giunto
joint: stopper16
Produttore/marchio commerciale
Corning 7661-16
Descrizione generale
Ground zone length indicates its greatest diameter (mm).
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.