CLS70581500
Pyrex® VISTA™ volumetric flask, class B
capacity 500 mL
Sinonimo/i:
flasks glass, volumetric flasks, volumetric glass
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass
long
Sterilità
non-sterile
Caratteristiche
closure type Snap cap
Confezionamento
case of 12
pack of 6
Produttore/marchio commerciale
Corning 70581-500
Capacità
500 mL
Descrizione generale
These Pyrex VISTA Class B volumetric flasks have capacity tolerances that are twice those specified for Class A volumetric ware. The graduation line is sharp and permanent; large blue clock letters make inscriptions easy to read. Snap-caps are supplied with all sizes.
Note legali
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)