Documenti fondamentali
CLS6706
Corning® LSE Platform Rocker
Double Platform, AC/DC input 230 V, AC/DC input 50/60 Hz, AC/DC input, Schuko plug
Sinonimo/i:
equipment replacement accessory
Scegli un formato
1 EA
CHF 1’170.00
CHF 1’170.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 1’170.00
About This Item
Codice UNSPSC:
41103811
NACRES:
NB.13
CHF 1’170.00
Check Cart for Availability
Ingresso CA/CC
, Schuko plug
Caratteristiche
Double Platform
Produttore/marchio commerciale
Corning 6706
Descrizione generale
The Corning LSE Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications. The adjustable rocking speed and fixed 7 degree tilt makes it optimal for staining fragile gels, as well as the more intensive action necessary for washing blots.
The speed of the Corning Platform Rocker can be adjusted from 2 to 60 rpm and the timer can be set for up to 2 hours or for continuous operation. The unit is safe to use in temperature controlled environments up to 70°C and is available in single or high capacity double platform configurations.
Features and Benefits
  • Low profile and compact size
  • 11-pound load capacity
  • Non-slip rubber pads to prevent sample containers from moving during operation
  • Double platform model available for increased capacity
Note legali
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)