CLS640660FO
PYREX® replacement Squibb separatory funnel w/o valve and stopper
pack of 4 ea
Sinonimo/i:
equipment replacement accessory
About This Item
Codice UNSPSC:
41121809
Confezionamento
pack of 4 ea
Produttore/marchio commerciale
Corning 6406-60FO
Capacità
60 mL
Volume
60 mL
Descrizione generale
These economy Squibb separatory funnel replacements do not have a Polytetrafluorethylene (PTFE) valve assembly or polyethylene stopper.
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)