Documenti fondamentali
CLS64062L
PYREX® economy Squibb separatory funnel
polyethylene stopper, PTFE stopcock, pack of 1 ea
Sinonimo/i:
pouring funnel
About This Item
Codice UNSPSC:
41121809
Materiali
PTFE stopcock
polyethylene stopper
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Corning 6406-2L
Alt. × diam. est. corpo
482 mm × 152.4 mm
Capacità
2 L
Lungh. stelo × D.E.
52 mm × 7 mm
Volume
2 L
20000 mL
Categorie correlate
Descrizione generale
These economical separatory funnels are designed to reduce replacement costs by providing replaceable components. The sturdy, durable funnels are designed for long life and maximum safety. The replaceable stopcock assembly has a simple screw thread locking nut and collar which ensure that the stopcock cannot fall out in use. The tip has a 7mm O.D. stem that is also replaceable. Its ground end provides a better grip and helps prevent the tip from falling out.
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)