CLS64061LFO
PYREX® replacement Squibb separatory funnel w/o valve and stopper
pack of 1 ea
Sinonimo/i:
equipment replacement accessory
About This Item
Codice UNSPSC:
41121809
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Corning 6406-1LFO
Capacità
1 L
Volume
1 L
1000 mL
Categorie correlate
Descrizione generale
These economy Squibb separatory funnel replacements do not have a Polytetrafluorethylene (PTFE) valve assembly or polyethylene stopper.
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)