Documenti fondamentali
CLS4931
Costar® vacuum aspirator
8-channel adapter
Sinonimo/i:
Costar aspirator, Vacuum aspirator
Scegli un formato
1 EA
CHF 308.00
CHF 308.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 308.00
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.11
CHF 308.00
Check Cart for Availability
Confezionamento
pack of 1
Produttore/marchio commerciale
Corning 4931
Categorie correlate
Descrizione generale
This is an 8-channel adapter that fits on to the Costar® Aspirator (CLS4930) so that it can be used to remove media and solutions from 8 wells simultaneously on a 96 well plate.
Please note: Only the 8-channel adapter is supplied for product CLS4931, despite the image on this web page showing both the adapter and the Costar® Aspirator (CLS4930) together they are sold separately.
Note legali
Costar is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)