Documenti fondamentali
CLS431840
Corning® centrifuge bottle
with silicon O-ring screw cap, capacity 250 mL, polypropylene
Sinonimo/i:
centrifugation bottles, digestion bottles, shaking bottles
Scegli un formato
36 EA
CHF 700.00
CHF 700.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
36 EA
CHF 700.00
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.11
CHF 700.00
Check Cart for Availability
Materiali
flat bottom tube
polypropylene
round bottle
translucent bottle
wide-mouth bottle
Sterilità
non-sterile
Caratteristiche
autoclavable
cap
Confezionamento
pkg of 36 ea
Produttore/marchio commerciale
Corning 431840
Parametri
27500 × g max. RCF
Capacità
250 mL
Caratteristiche e vantaggi
  • Corning® centrifuge bottles have a specially designed cap for improved grip
  • Includes clear graduations and textured marking spot.
  • Bottles are designed with extra-thick walls for high durability
  • Wide mouth for easier liquid filling
  • Compatible with most centrifuges and do not require special buckets
  • Unique design aid researchers to reduce the number of centrifugation cycles and enhance productivity
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)