Documenti fondamentali
CLS11500
Corning® Transfer Cap with Diptube for 5 L Erlenmeyer Flasks
polypropylene, sterile, case of 4 ea
Sinonimo/i:
Cap for 5 L Flasks, Corning Transfer Cap, Transfer Cap with Diptube
Scegli un formato
4 EA
CHF 775.00
CHF 775.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
4 EA
CHF 775.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.11
CHF 775.00
Check Cart for Availability
Materiali
orange
polypropylene
Sterilità
sterile
Confezionamento
case of 4 ea
Produttore/marchio commerciale
Corning 11500
Diam. tappo
100 mm
Categorie correlate
Descrizione generale
Closed cell culture systems help reduce the risk of adventitious contamination. Corning offers closed systems accessories with vessels to help transfer liquid aseptically. The internal end is a rigid polypropylene dip tube designed to reach the bottom of the flask. The venting port ends in a 50 mm diameter vent filter with a 0.2 μm pore PVDF membrane and a barbed fitting. The liquid transfer port uses a 1/4" ID (3/8" OD) chemically resistant, heat sealable flexible tubing on the external side of the cap ending in a male plastic coupler (MPC) male disconnect with a sealing cap.
Caratteristiche e vantaggi
  • Animal component free vessels
  • SAL 10-6 sterilization
  • Wide range of product offerings
  • Customizable solutions
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)