Documenti fondamentali
AXYVT250RLAB
Pipette tips for Velocity, robotic
size 250 μL, pkg of 5x960tips/cs, racked in bottom sealed rack
Sinonimo/i:
Pipette tips for Velocity
About This Item
Codice UNSPSC:
41121600
NACRES:
NB.02
Prezzi e disponibilità al momento non sono disponibili
Sterilità
non-sterile
Produttore/marchio commerciale
Axygen Inc VT-250-R-LAB
Corning
Confezionamento
pkg of 5x960tips/cs
Misura
250 μL
Volume
250 μL
Caratteristiche e vantaggi
Axygen® 96-well tips have the following features and benefits:
  • For greater consistency and performance they are assembled via automation
  • Tips are RNAse/DNAse free
  • Tips are made of polypropylene and are non-pyrogenic
  • Compatible with Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST and 16ST head
Altre note
Browse all pipette tips
Note legali
Agilent is a registered trademark of Agilent Technologies, Inc.
Axygen is a registered trademark of Corning, Inc.
Velocity11 VPrep is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)