Documenti fondamentali
AXYPCR0208C100
Corning® Axygen® PCR Tube Strips
Sinonimo/i:
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
Scegli un formato
10 X 125 STRIPS
CHF 154.00
CHF 154.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 X 125 STRIPS
CHF 154.00
About This Item
Codice UNSPSC:
24122002
NACRES:
NB.22
CHF 154.00
Check Cart for Availability
Produttore/marchio commerciale
Corning PCR-0208-C-100
Descrizione generale
  • Corning® Axygen® PCR tube strips are made of 99.9% pure virgin polypropylene.
  • Designed to fit a wide variety of thermal cyclers.
  • Thin, consistent, and uniform wall thickness facilitate accurate thermal transfer and optimal amplification.
  • An even insertion cap prevents evaporation and cross-contamination of the samples.
  • Tubes are free of RNase-/DNase and nonpyrogenic.
Note legali
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Articoli
Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.