Documenti
AXYP384240SQCSI
Corning® Axygen® Deep Well Microplate
indiv. wrapped, size 384 wells, working volume 240 μL, pkg of 50 ea, sterile
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Materiali
square bottom clear polypropylene wells
Sterilità
sterile
Confezionamento
pkg of 50 ea
Produttore/marchio commerciale
Axygen Inc P-384-240SQ-C-SI
Corning
Volume massimo
250 μL
Misura
384 wells
Volume d’esercizio
240 μL
Tipo di binding
non-treated surface
Note legali
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.