Documenti
Z800864
Hellma® fluorescence cuvettes, sealable cells
Suprasil® quartz, spectral range 200-2500 nm, pathlength 10x10 mm, chamber volume 3500 μL
About This Item
Codice UNSPSC:
12352200
Materiali
Suprasil® quartz
Descrizione
4 windows
Produttore/marchio commerciale
Hellma 117.200F-QS
Parametri
200-2500 nm spectral range
Alt. × largh. × prof.
56 mm × 12.5 mm × 12.5 mm
Volume della camera
3500 μL
Cammino ottico
10x10 mm
Descrizione generale
macro, semi-micro, for anaerobic applications
Note legali
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Choose from one of the most recent versions:
Certificati d'analisi (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Servizio Clienti