CFO09NC
Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids
Size 400, Nom. Constant 1.2, Range 240 to 1,200 cSt, (non certified)
About This Item
Codice UNSPSC:
41116107
NACRES:
NA.24
Produttore/marchio commerciale
Paragon Scientific Ltd
applicazioni
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Descrizione generale
This is the non certified version of this viscometer. If you wish to order a certifed viscometer, please refer to the corresponding product CFO09UKC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)