PA38/111
NovAseptic® Valve, Pneumatic Plastic Actuator, Normally Closed, NC
About This Item
Codice UNSPSC:
40141600
eCl@ss:
37051001
Prodotti consigliati
Materiali
POM-C indicator
cover (Stainless Steel EN 1.4305/ASTM 303 and Plastic POM C)
Livello qualitativo
Nome Commerciale
NovAseptic®
Parametri
pressure range at -1--6 bar
temp. range at 0-70 °C
Peso
2.2 kg (4.85 lb)
Applicazioni
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Altre note
Actuator, Pneumatic
Note legali
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany