Vai a
MXQULTVP2
Milliflex® Quantum system Validation Protocol for the combination with Milliflex® Oasis system
for use with Milliflex® Quantum Systems, for use with Milliflex Oasis® Systems
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
A voi/SKUDisponibilitàPrezzo
Informazioni su questo articolo
NACRES:
ND.05
UNSPSC Code:
41116500
Prezzi e disponibilità al momento non sono disponibili
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
availability
available only in North America
compatibility
for use with Milliflex Oasis® Systems, for use with Milliflex® Quantum Systems
General description
Milliflex® Quantum Validation Protocol(Letter Format) is designed as a ready to use step by step guide to perform IQ and OQ activities of rapid microbial detection system using Milliflex® Quantum in combination with Milliflex®Oasis system. The protocol contains an exhaustive list of tests including test descriptions and test records. The protocol follows GMP, EP, USP and JP regulatory guidelines. Only available in North America.
Application
Milliflex® Quantum Validation Protocol is used to validate your rapid microbial detection system with ease and consistent performance while following regulatory guidelines.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
MXQUA4VP2RPSTA4VP1MAVFA4VP2
compatibility
for use with Milliflex Oasis® Systems, for use with Milliflex® Quantum Systems
compatibility
for use with Milliflex Oasis® Systems, for use with Milliflex® Quantum Systems
compatibility
for use with Milliflex® Rapid Systems.
compatibility
for use with MAS-100 VF® Microbial Air Samplers
availability
available only in North America
availability
-
availability
-
availability