MP0DDSPMAN3
Proprietà
hydrophilic
300
pkg of 3 racks
Millistak+®
liquid
Descrizione
Mostra le differenze
1 of 1
Millipore
Millistak+® Pod Disposable Manifold
MP0DDSPMAN2
Millistak+® Pod Manifold
MCE30027H1
Filtro di profondità Millistak+® Pod
MCE35027H1
CE30, cellulose, filtration area 0.027 m2, hydrophilic
CE35, cellulose, filtration area 0.027 m2, hydrophilic
sample type liquid
-
400
Explore all of our products under Millistak+® Pod Disposable Manifold
Scegli una delle versioni più recenti:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documenti section.