Documenti fondamentali
Z763241
CRC Handbook of Chemistry and Physics, 93rd Edition
Haynes, William M., ed., CRC Press, 2012, 2664 pp., hard cover
About This Item
Codice UNSPSC:
55101509
Info sulla pubblicazione
Haynes, William M., ed., CRC Press, 2012, 2664 pp., hard cover
Produttore/marchio commerciale
CRC Press
Indice
Basic Constants, Units, and Conversion Factors. Symbols, Terminology, and Nomenclature. Physical Constants of Organic Compounds. Properties of the Elements and Inorganic Compounds. Thermochemistry, Electrochemistry, and Kinetics. Fluid Properties. Biochemistry. Analytical Chemistry. Molecular Structure and Spectroscopy. Atomic, Molecular, and Optical Physics. Nuclear and Particle Physics. Properties of Solids. Polymer Properties. Geophysics, Astronomy, and Acoustics. Practical Laboratory Data. Health and Safety Information. APPENDICES. Mathematical Tables. Sources of Physical and Chemical Data. Index.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)