Documenti fondamentali
Z743937
Ohaus® Explorer® semi-micro analytical balance
Model EX125DM, weighing capacity 52 or 120 g, Precision 0.01 0.1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Sinonimo/i:
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
Scegli un formato
1 EA
CHF 8’700.00
CHF 8’700.00
Spedizione prevista il18 maggio 2025
Scegli un formato
Cambia visualizzazione
1 EA
CHF 8’700.00
About This Item
Codice UNSPSC:
41111502
NACRES:
NB.71
CHF 8’700.00
Spedizione prevista il18 maggio 2025
Materiali
stainless steel weighing plate
Caratteristiche
Model EX125DM
Produttore/marchio commerciale
Ohaus® 30139515
Precision measuring range
0.01 0.1 mg
Capacità di pesata
52 or 120 g
Diam. piattaforma
3.14 in. (80 mm)
Descrizione generale
Applications
Weighing, Percent Weighing, Parts Counting, Check Weighing, Dynamic/Animal Weighing, Filling, Totalization, Formulation, Differential Weighing, Density Determination, Peak Hold, Pipette Adjustment, SQC
Display
Full-color VGA graphic display (5.7 in. diagonal), 4-wire resistive touch screen. QWERTY keyboard and numeric keypad to quickly input GLP and GMP data and other application data
Operation
AC Adapter (Included)
Communication
Easy access communication ports including 2 units of USB port and RS232 and an optional fourth port of either RS232 or Ethernet. GLP/GMP Data Ouput with Real Time Clock. Data Transfer Function.
Construction
Metal base, ABS top housing, stainless steel pan,anti-static glass draftshield with flip-top door, replaceable in-use cover, automatic door as option models
Design Features
Fully-automatic internal calibration system, fast stabilization time, four touchless sensors, built-in ionizer with automatic door models, up to 14 operating languages, OIML models available, SAVE to USB function, menu lock switch, security bracket, integral weigh below hook for below balance weighing applications, removable stainless steel weighing platform, stability indicator, overload and underload indicators, auto standby.
Note legali
Explorer is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)