Documenti fondamentali
Z742419
Q-Tube gas purging set
12 SS, pack of 1 ea
Scegli un formato
1 EA
CHF 1’890.00
CHF 1’890.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 1’890.00
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.51
CHF 1’890.00
Check Cart for Availability
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Q Labtech 100-QT00PG-012SS
Parametri
180 psi max. pressure
Descrizione generale
The parts with direct chemical contact are made of 316 stainless steel, PTFE, Simriz, or glass.
Q-Tube-Purging-12-SS / 100-QT00PG-012SS can be operated safely and accurately with high reproducibility with a wide range of gases, including HCl, NH3, H2, O2, N2, Ar, D2, CO, CO2, and etc. The set contains all items needed for immediate use.
Note legali
Q-Tube is a trademark of Q Labtech LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)