Documenti
Z693766
LabJaws lattice rod end connector
nickel plated
Sinonimo/i:
clamp holders, frame connectors
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.35
Materiali
nickel plated
Produttore/marchio commerciale
Troemner 916126
Capacità ganasce
0-13 mm
Descrizione generale
Holds rods firmly at 90°. Use when semi-permanent installations are required. Comes with two set screws.
Note legali
LabJaws is a trademark of Henry Troemner LLC