Z691143
Clipox™ ground glass joint clip for conical joints
size NS60
Sinonimo/i:
clamp clips, glass clips, joint clips, steel clamps
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
Materiali
stainless steel
Misura
NS60
Produttore/marchio commerciale
Clipox 10060
Descrizione generale
Clipox are safe, universal clips for ground joints manufactured from high grade stainless steel. Their slim, symmetric design solves application problems of common clips and clamps. They are more secure, have better thermal and chemical resistance and are able to be used with stand clamps.
Clipox are available to secure conical, spherical, stopcocks and glass stopper ground joints.
Clipox are available to secure conical, spherical, stopcocks and glass stopper ground joints.
- Increases the safety of glass instruments
- Secure horizontal and vertical in case of pressure overload
- Easy to mount and to remove due to two-sided handles
- Longevity of suspension and shape due to spring steel wire
- Outstanding thermal, chemical and acid resistance
- Reduced scratch of glass surface due to polished surface of material
- Made in Germany, production certified according to ISO 9001
Note legali
Clipox is a trademark of Clipox Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)