Documenti fondamentali
Z687944
Levochem ethylene-vinyl-alcohol gloves
size 10
About This Item
Codice UNSPSC:
46181504
Misura
10
Caratteristiche
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
Produttore/marchio commerciale
KCL 761-10
Confezionamento
pkg of Packaged in boxes of 10 pairs and cases of 10 boxes
Lungh. × spessore
360-400 mm × 0.07 mm , +/- 0.01 mm
Descrizione generale
Levochem is made of ethylene-vinyl-alcohol (EVOH) and polyethylene welded 3-layer sandwich system. It offers high resistance against a variety of hazardous substance groups and can be worn on its own or under another glove.
  • Very good protection against variety of hazardous substance groups
  • Can be worn under gloves
  • Excellent for use in emergency spill applications
  • Ambidextrous
  • CE compliant
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)