Z682322
Ace rotary evaporator condenser complete "A" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144, includes 1 L receiving flask and clamp
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
plastic-coated glass
Produttore/marchio commerciale
Büchi equivalent, 40632
Descrizione generale
Includes 1 L receiving flask and clamp.
The complete "A" evaporator condenser assembly includes the main glass condenser body with inner coil for angled installation, GL threaded caps and hose barbs. Bottom joint is 35/20. It is designed to fit Büchi® evaporator models 200/205 and series 114-144.
Note legali
Büchi is a registered trademark of Büchi Labortechnik AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)