Documenti fondamentali
Z680559
Caps for Duran® laboratory bottles
neck thread for GL 45, blue-yellow polypropylene tamper-evident (with lip seal)
Sinonimo/i:
caps bottles, glass bottles caps
Scegli un formato
10 EA
CHF 72.00
CHF 72.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 EA
CHF 72.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
CHF 72.00
Check Cart for Availability
Produttore/marchio commerciale
DURAN®
Schott
Raccordi
neck thread for GL 45
Descrizione generale
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)