Merck
Tutte le immagini(5)
Documenti fondamentali
Z680494
Duran® GL 45 connection system caps and accessories
insert for hose connection screw cap GL 14, 6.0 mm
Sinonimo/i:
1129818, Duran Cap Accessories, GL 45 Connection Caps
Scegli un formato
1 EA
CHF 15.80
CHF 15.80
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 15.80
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
CHF 15.80
Check Cart for Availability
Materiali
white PTFE/silicone
Produttore/marchio commerciale
DURAN® 1129818
Parametri
-50-230 °C temp. limit
Descrizione generale
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Caratteristiche e vantaggi
  • Manufactured from PP, PTFE, silicone
  • Accessories designed for use with the DURAN® GL 45 2/3 port tubing connection caps or GL 45 / GLS 80® Stirred Bottle Reactor caps
  • Safe and secure liquid transfer into or out of laboratory bottles
  • Four inserts are suitable for rigid or semi-rigid tubing with a range of outer diameters: 1.6, 3.0, 3.2 or 6.0 mm
  • Open-topped blue PP GL 14 pass-through tubing cap for use with the inserts
  • Closed-top red PBT GL 14 cap for sealing unused ports
  • Sterile pressure equilisation is possible with pressure compensation set
Note legali
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)