Documenti fondamentali
Z678899
FORTUNA® Universal Optifix® HF dispenser
dispensing volume 10-50 mL
Scegli un formato
1 EA
CHF 2’150.00
CHF 2’150.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 2’150.00
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.01
CHF 2’150.00
Check Cart for Availability
Produttore/marchio commerciale
Fortuna Graf 101.106-48
dispensing volume measuring range
10-50 mL
Volume
10-50 mL
Raccordi
GL 40
GL 45
Categorie correlate
Descrizione generale
For all liquids that can be handled with the Optifix dispensers and for hydrofluoric acid (HF)
Features:
  • Dispensing cylinder made of PTFE
  • Massive PTFE encased dispensing piston
  • FIX adapter made of PTFE
  • Ergonomic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
  • Valve block made of PTFE, with additional safety valve in the ventilation tube, no metal springs in the valves - no corrosion
Supplied:
FORTUNA Universal Optifix HF dispenser, with 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
Note legali
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)