Documenti fondamentali
Z677590
Camatril nitrile long cuff gloves
size M
Sinonimo/i:
disposable glove
About This Item
Codice UNSPSC:
46181504
NACRES:
NB.35
Materiali
green gloves
green nitrile
Misura
M
Sterilità
non-sterile
Caratteristiche
CE compliant
disposable
latex free
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
Confezionamento
pack of 1 pair
Produttore/marchio commerciale
KCL 733
KCL 733
Lungh. × spessore
600 mm × 0.5 mm
Descrizione generale
Camatril nitrile long cuff gloves are of high standard, with a wide range of applications and made of specific high quality Nitrile. The large spectrum of chemical resistance ensures the use of these gloves in numerous fields. The remarkably high abrasion, cut and puncture resistance allows work with sharp edged materials. Good flexibility, anatomical design and excellent fit ensure a good sense of touch.
  • Ergonomic fit and high elasticity ensures good sensitivity
  • Roughened palm for firm grip when wet
  • High mechanical resistance
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • CE compliant
Note legali
Camatril is a trademark of KCL GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)