Documenti fondamentali
Z671932
KNF LIQUIPORT chemically-resistant diaphragm liquid pumps
AC/DC input 115 V AC, polypropylene (head), flow rate 0.5-3.0 L/min
About This Item
Codice UNSPSC:
41105101
NACRES:
NB.79
Materiali
polypropylene (head)
Caratteristiche
CSA approved
Produttore/marchio commerciale
KNF NF 300 KT.18 S
Disponibilità
not available in EU
Parametri
0.5-3.0 L/min flow rate
D.I. tubo
12 mm
Peso
1.5 kg
Uscita
24 watts (motor)
Descrizione generale
KNF′s new NF liquid pumps are the convenient way to transfer liquids in the laboratory without the mess of peristaltic tubing. They are stylish and compact, and PTRFE models are highly resistant to chemical attack, prevent contamination f your pumped medium.
The NF100 and NF300 pumps are available in either polypropylene or PTFE head materials, with FFPM valve. You should always check corrosion compatibility charts with the medium you intend to pump.
  • 10 mWg pressure head
  • 3 mWg suction head
  • IP65 protection class
  • Maintenance-free
  • Self priming
  • Dry running
  • Adjustable flow rate
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)