Documenti fondamentali
Z648477
Quickfit® multi-limb vacuum receiver
Quickfit RA 11/SC, four-limb vacuum receiver
Sinonimo/i:
RA11/SC
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.43
Giunto
female joint 14/23
male joint 19/26
Produttore/marchio commerciale
Quickfit RA 11/SC
Disponibilità
not available in ROW (Available only in EU countries)
not available in USA and Canada
Descrizione generale
When suitably lubricated the receiver adapter may be rotated at the conical joint to divert liquid, in turn, to four attached flasks without disturbing the vacuum in the system. Requires four 100 mL flasks (Z302619, not included) with screw-thread adapter for flexible tubing.
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)