Documenti fondamentali
Z645265
IKA® sieves for MF 10 basic microfine grinder drive
MF 0.25, 0.25mm, 1/cs
Scegli un formato
1 EA
CHF 550.00
CHF 550.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 550.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.74
CHF 550.00
Check Cart for Availability
Confezionamento
case of 1
pack of 1
Produttore/marchio commerciale
IKA 2938900
Misura
0.25 mm
Categorie correlate
Descrizione generale
Note: Interchangeable sieve for insertion into the grinding heads ensures maximum particle size filtering.
Note legali
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Alexey Veremeev et al.
Materials (Basel, Switzerland), 13(15) (2020-08-06)
Here we evaluated the efficacy of bone repair using various native bovine biomaterials (refined hydroxyapatite (HA), demineralised bone matrix (DBM), and purified bone collagen (COLL)) as compared with commercially available bone mineral and bone autografts. We employed a conventional critical-sized