Documenti
Z602876
Normag reflux boiling splitter
bottom joint: 29/32
About This Item
Codice UNSPSC:
12352200
NACRES:
NB.43
Giunto
bottom joint: 29/32
side joint 14/23
Produttore/marchio commerciale
Normag
Disponibilità
not available in USA and Canada
Caratteristiche e vantaggi
With fine adjustment glass needle valve. Installation length 50mm for
thermometers. GL 14 Glass thread cooling water connections. Complete
with 2 GL 14 threaded hose connectors.
Note legali
Normag is a trademark of Normag Labor- und Prozesstechnik GmbH