Documenti fondamentali
Z551694
Chemat vacuum chucks
diam. 1 1/2 in.
Scegli un formato
1 EA
CHF 469.00
CHF 469.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 469.00
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.79
CHF 469.00
Check Cart for Availability
Descrizione
Type CG
Produttore/marchio commerciale
Chemat
Diametro
1 1/2 in.
Descrizione generale
For use with the Chemat Spin-coater system.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Questions
  1. Greetings! We are interested to buy a vacuum chunk of model Z551694-1EA for our spin coater. To proceed with that we need to know the size of hole in the chunk. Could you please inform us about the details of the chunk,specially hole diameter ?
    1 answer
    1. As mentioned on the Product Detail Page in the PROPERTIES section, the diameter is given as 1 1/2 inches.