Documenti fondamentali
Z549223
Sigma-Aldrich
Aldrich® antisplash splash-guard adapter with return hole
female joint: ST/NS 29/32, male joint: ST/NS 14/20, capacity 100 mL
Scegli un formato
1 EA
CHF 165.00
CHF 165.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 165.00
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
CHF 165.00
Check Cart for Availability
Giunto
female joint: ST/NS29/32
male joint: ST/NS14/20
Capacità
100 mL
Categorie correlate
Descrizione generale
These traps greatly reduce carry-over from foaming or bumping. Anything that may climb can return back down through the return hole at the bottom of this adapter.
Applicazioni
Aldrich antisplash splash-guard adapter with return hole may be used between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into the condenser in case of "bumping". A drain hole at the base of the inner tube allows solvent to drain back into the evaporation flask.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)