Documenti fondamentali
Z549053
Sigma-Aldrich
Aldrich® anticlimb splash-guard adapter
female joint: ST/NS 29/32, male joint: ST/NS 24/40, capacity 100 mL
Scegli un formato
1 EA
CHF 188.00
CHF 188.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 188.00
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
CHF 188.00
Check Cart for Availability
Giunto
female joint: ST/NS29/32
male joint: ST/NS24/40
Capacità
100 mL
Categorie correlate
Descrizione generale
Aldrich® anticlimb splash-guard adapter greatly lower carry-over from foaming or bumping. Because the venting hole is located at the top, the foam can be prevented from climbing higher into the condenser.
Applicazioni
Aldrich® anticlimb splash-guard adapter are ideal for use with solvents that foam excessively during rotary evaporation. Inner vapor tube is sealed to the top of trap to prevent foam entry into condenser.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)