Z530689
Aldrich® vacuum manifold with joints
positions, 5, joint: ST/NS 14/20
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.79
Prodotti consigliati
Giunto
joint: ST/NS14/20
Caratteristiche
positions 5
Lunghezza
500 mm
Categorie correlate
Descrizione generale
This practical design provides clearance to accommodate 250mL flasks with a snap of a Keck® clip. Manifolds are available with either <ST/>14/20, <ST/>24/40, or <ST/>29/32 joints. Glass stopcocks have a 4mm bore.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)