Documenti
Z530565
Sigma-Aldrich
Aldrich® reagent bottle
joint: ST/NS 14/20, capacity 60 mL, amber bottle
Sinonimo/i:
bottle reagent, reagent bottles
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
amber bottle
glass bottle
glass cap
round bottle
Giunto
joint: ST/NS14/20
Caratteristiche
with cap
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Sigma-Aldrich
Capacità
60 mL
Descrizione generale
Reagent bottles come with a ground glass hex head stopper. For light-sensitive samples, use the amber coated reagent bottles.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC