Documenti fondamentali
Z404624
Sigma-Aldrich
Aldrich® splash protector
female joint: ST/NS 29/32, male joint: ST/NS 29/32
Sinonimo/i:
Laboratory Splash Shield
Scegli un formato
1 EA
CHF 159.00
CHF 159.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 159.00
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
CHF 159.00
Check Cart for Availability
Giunto
female joint: ST/NS29/32
male joint: ST/NS29/32
Descrizione generale
Effectively prevents upward movement of foaming or bumping liquids in rotary evaporator traps. Unique design has two baffles with horizontally opposing holes that may also be used with other distillation techniques.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)