Documenti fondamentali
Z304581
Quickfit® ground-glass stoppers
joint: ST/NS 10/19
Sinonimo/i:
SB10
Scegli un formato
1 EA
CHF 12.70
CHF 12.70
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 12.70
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.35
CHF 12.70
Check Cart for Availability
Giunto
joint: ST/NS10/19
Produttore/marchio commerciale
Quickfit SB 10
Disponibilità
not available in USA and Canada
Descrizione generale
  • Ideally suited for use with Quickfit® jointedware and other glassware having standard ground glass sockets
  • All Quickfit® stoppers conform to the standard tapers as outlined in ISO 383
  • Borosilicate glass for chemical durability and high temperature performance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)