Documenti fondamentali
Z303917
Quickfit® flat flange lids for reaction flasks
center joint: ST/NS 34/35, parallel socket joint: 19/26, 10° side socket Joints: 19/26 (2)
Sinonimo/i:
MAF5/2
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
borosilicate glass
Etichetta
B
Giunto
10° side socket Joints: 19/26 (2)
15° side socket joint
5° side socket joint
center joint: ST/NS34/35
parallel socket joint: 19/26
Sterilità
non-sterile
Confezionamento
pack of 1
Produttore/marchio commerciale
Quickfit MAF 5/2
Disponibilità
not available in USA and Canada
Descrizione generale
Single and multi-socket lids for reaction flasks for simple extraction, reflux or for the introduction of a variety of stirrers, dropping funnels, temperature probes etc.
Use clip to secure lid to flasks, purchase separately.
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)