Documenti fondamentali
Z261483
Luer-to-Tubing Connector
bulkhead panel mounted Micro-Mate® female Luer to hose end for 1/8 in. to 3/16 in. I.D. tubing, nickel plated
Sinonimo/i:
environmental, veterinary
Scegli un formato
1 EA
CHF 56.30
CHF 56.30
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 56.30
About This Item
Codice UNSPSC:
40183110
NACRES:
NB.03
CHF 56.30
Check Cart for Availability
Materiali
nickel plated
Produttore/marchio commerciale
Popper 6420
Categorie correlate
Descrizione generale
Hardware specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating, easy to clean and resistant to corrosion.
Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Note legali
Micro-Mate is a registered trademark of Cadence Science
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)