Documenti fondamentali
Z258415
Sigma-Aldrich
Vortex-Genie® 2 mixer
Vortex-Genie® 2 mixer
AC/DC input 120 V AC
Sinonimo/i:
Genie Mixer, Vortex Mixer
About This Item
Codice UNSPSC:
41103800
NACRES:
NB.73
Prezzi e disponibilità al momento non sono disponibili
Ingresso CA/CC
120 V
Caratteristiche
CE compliant
CSA approved
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Scientific Industries SI-0236
Scientific Industries
Disponibilità
not available in EU
Descrizione generale
Mixes liquids quickly and thoroughly with vortex action. Variable speed control allows precise mixing range. The unit contains a standard 3 in. platform head for flasks and beakers, and a cup head for single tube mixing. Accessories for special applications are sold separately. Speed: 600 to 3,200 rpm. Max. load: 1 lb.
  • Variable speed control
  • Shaking or vortex action
  • Continuous or touch-control mixing
Accessories sold separately
Note legali
Vortex-Genie is a registered trademark of Scientific Industries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
Questions
  1. Is it possible to clarify which accessories are included with Z258415-1EA, as the picture on the website is confusing?
    1 answer
    1. No accessories are included, only the vortexer itself.
      Helpful?
Reviews
Active Filters
  1. 1 Ratings-Only Review