Documenti fondamentali
Z256331
Wrap-it ties
L 7 1/2 in.
Sinonimo/i:
tie wrap, wrap tie
Scegli un formato
250 EA
CHF 56.30
CHF 56.30
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
250 EA
CHF 56.30
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.35
CHF 56.30
Check Cart for Availability
Parametri
50 lb tensile strength
Lunghezza
7 1/2 in.
Descrizione generale
Self-locking, one-piece nylon construction. Excellent for securing rubber septa to bottles and flask joints, attachment of tubing to hose barbs or glassware, and for closing plastic bags or drum liners. Use with Wrap-it tie installing tool.
Tear-off version is blue in color and is easily removed from septum with a quick pulling motion.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)