Documenti fondamentali
Z243256
ACE Squibb separatory funnel with Locktight stopper
capacity 60 mL, stopper joint: ST/NS 24/40
Scegli un formato
1 EA
CHF 278.00
CHF 278.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 278.00
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
CHF 278.00
Check Cart for Availability
Giunto
joint: ST/NS stopper24/40
Produttore/marchio commerciale
Ace Glass 723155
Capacità
60 mL
Categorie correlate
Descrizione generale
European style funnel with new leaktight locking stopper that will not fall out during shaking or transportation. The stopper is secured into a screw-threaded top joint via cap and Viton® O-ring for a positive, leaktight seal. Supplied complete with 1:5 PTFE plug, stopper, cap, and O-ring.
Note legali
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)