Documenti fondamentali
Z242772
Scienceware® Vikem® vinyl-coated lead wire
wire diam. 1/4 in.
Sinonimo/i:
coil wire, wire spiral, wire vinyl
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.43
Materiali
lead (coated)
Sterilità
non-sterile
Confezionamento
pack of 1
Produttore/marchio commerciale
Bel-Art F18324-0000
Diam. filo
1/4 in.
Peso
114 g
Descrizione generale
Can be coiled into a helix, spiral, cone, or any irregular shape to fit around flasks and bottle necks to stabilize them in a water bath. 12 in. length. Coated with chemically-resistant red Vikem vinyl.
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vikem is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)