As listed on the Product Detail Page in the PROPERTIES section, the product dimensions are 4 in. × 2 7/16 in. ( I.D. × H.). The weight of the desiccator is 19oz.
Z225096
Scienceware® mini-vacuum desiccator
pkg of 1 ea
Scegli un formato
CHF 138.00
Scegli un formato
About This Item
CHF 138.00
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bel-Art F41099-0000
Diam. int. × alt.
4 in. × 2 7/16 in.
Descrizione generale
Not compatible with organic solvents and vapors.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
-
What's the internal diameter of the dessiccator?
1 answer-
Helpful?
-
-
Can the product Z225096 be subjected to a temperature of 140°C for a duration of 2 hours?
1 answer-
The Scienceware mini-vacuum desiccator Z225096 cannot be heated as it would melt, according to the supplier.
Helpful?
-
-
Do the Scienceware® brand polypropylene products contain any leachable material?
1 answer-
Although Bel-Art does not specifically test their Scienceware® brand polypropylene products, they have no evidence of any problems with leachable material in the polypropylene.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
Potete fornirmi dimensioni e peso del mini essiccatore a vuoto?
0 answers