Documenti fondamentali
Z220108
Sigma-Aldrich
Aldrich® micro distillation head with Vigreux indents
One-piece assembly
Scegli un formato
1 EA
CHF 1’700.00
CHF 1’700.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 1’700.00
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.43
CHF 1’700.00
Check Cart for Availability
Categorie correlate
Descrizione generale
Has a vacuum-jacketed head with attached condenser, Vigreux indents and thermowell for 50 mm immersion thermometer. Joints on the one-piece are 14/20 inner joints and 10/18 outer joints. Vigreux indents run the entire length of the column and act as the packing for the column.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)